Gene Transcripts

Gene Symbol
OR52N2
Entrez Gene ID
390077
Gene Name
olfactory receptor family 52 subfamily N member 2
Gene Aliases
OR11-57
Chromosome Location
Chr. 11: 5803433 - 5822675 on Build GRCh38
UniGene ID
Hs.553737
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005174.1 NP_001005174.1 1 41
XM_011520090.2 XP_011518392.1 2 5717
GenBank mRNA BC140732.1 - 1 63

View Full Product Details