Gene Transcripts

Gene Symbol
MYL9
Entrez Gene ID
10398
Gene Name
myosin light chain 9
Gene Aliases
LC20, MLC-2C, MLC2, MRLC1, MYRL2
Chromosome Location
Chr. 20: 36541484 - 36549823 on Build GRCh38
UniGene ID
Hs.504687
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006097.4 NP_006088.2 4 1176
NM_181526.2 NP_852667.1 3 1014
GenBank mRNA AF176042.1 - 3 1113
AF176043.1 - 2 951
AK130879.1 - 4 1141
BC002648.2 - 3 953 3608316
D82057.1 - 4 1145
J02854.1 - 3 1098

View Full Product Details