Gene Transcripts

Gene Symbol
MDM1
Entrez Gene ID
56890
Gene Name
Mdm1 nuclear protein
Gene Aliases
Chromosome Location
Chr. 12: 68294566 - 68332381 on Build GRCh38
UniGene ID
Hs.655702
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001205029.1 NP_001191958.1 3 2294
NM_017440.4 NP_059136.2 4 724
NM_020128.2 NP_064513.1 3 1805
XM_005269026.1 XP_005269083.1 4 649
XM_011538569.1 XP_011536871.1 4 649
XM_017019646.1 XP_016875135.1 3 1726
XM_017019647.1 XP_016875136.1 4 649
XM_017019648.1 XP_016875137.1 4 1156
XM_017019650.1 XP_016875139.1 3 2237
GenBank mRNA AF267851.1 - 3 1727
BC028355.2 - 4 675 4827410
BC034945.1 - 3 269

View Full Product Details