Gene Transcripts

Gene Symbol
C22orf29
Entrez Gene ID
79680
Gene Name
chromosome 22 open reading frame 29
Gene Aliases
BOP
Chromosome Location
Chr. 22: 19846138 - 19854848 on Build GRCh38
UniGene ID
Hs.105642
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024627.5 NP_078903.3 3 1140
GenBank mRNA AK024778.1 - 1 516
AK225088.1 - 1 516
AK291262.1 - 3 1108
AL832509.1 - 3 1029
BC011679.2 - 1 647 4303449
BX640785.1 - 2 1229
BX640969.1 - 3 1109
BX640998.1 - 3 1109

View Full Product Details