Gene Transcripts
- Gene Symbol
- PTRH1
- Entrez Gene ID
- 138428
- Gene Name
- peptidyl-tRNA hydrolase 1 homolog
- Gene Aliases
- C9orf115, PTH1
- Chromosome Location
- Chr. 9: 127695024 - 127716714 on Build GRCh38
- UniGene ID
- Hs.653265
- Species
- Homo sapiens
- Gene Symbol
- CFAP157
- Entrez Gene ID
- 286207
- Gene Name
- cilia and flagella associated protein 157
- Gene Aliases
- C9orf117
- Chromosome Location
- Chr. 9: 127706261 - 127716002 on Build GRCh38
- UniGene ID
- Hs.652519
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL833241.1 |
- |
2 |
1113 |
|
View Full Product Details