Gene Transcripts

Gene Symbol
PRIMA1
Entrez Gene ID
145270
Gene Name
proline rich membrane anchor 1
Gene Aliases
PRIMA
Chromosome Location
Chr. 14: 93718298 - 93789029 on Build GRCh38
UniGene ID
Hs.432401
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178013.3 NP_821092.1 1, 2 135
XM_011536456.2 XP_011534758.1 2, 3 589
GenBank mRNA AY225516.1 - 1, 2 135
AY225517.1 - 1, 2 135

View Full Product Details