Gene Transcripts

Gene Symbol
THAP8
Entrez Gene ID
199745
Gene Name
THAP domain containing 8
Gene Aliases
Chromosome Location
Chr. 19: 36034984 - 36054785 on Build GRCh38
UniGene ID
Hs.350209
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152658.2 NP_689871.1 1, 2 617
GenBank mRNA AK057453.1 - 2, 3 473
AK093048.1 - 1, 2 618
BC046207.1 - 1, 2 166 6176484
BC072416.1 - 1, 2 257 6169558
JQ410992.1 - 1, 2 617

View Full Product Details