Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173545.2 | NP_775816.1 | 8 | 1374 | |
GenBank mRNA | AK290841.1 | - | 8 | 1371 | |
AK308186.1 | - | 7 | 1264 | ||
BC030711.2 | - | 4 | 570 | ||
BC041144.1 | - | 8 | 1323 | 6042653 |