Gene Transcripts

Gene Symbol
TBATA
Entrez Gene ID
219793
Gene Name
thymus, brain and testes associated
Gene Aliases
C10orf27, SPATIAL
Chromosome Location
Chr. 10: 70771237 - 70785419 on Build GRCh38
UniGene ID
Hs.386698
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK057382.1 - 8 1108
AK303344.1 - 7 797
AK310732.1 - 3 461
BC137543.1 - 8 1160 9021543
BC137544.1 - 8 1094 9021544
BC144692.1 - 8 1091 9053226
BC144694.1 - 8 1097 9053228
DB147708.1 - 5 541
HY259649.1 - 1 52

View Full Product Details