Gene Transcripts

Gene Symbol
MT1DP
Entrez Gene ID
326343
Gene Name
metallothionein 1D, pseudogene
Gene Aliases
MTM
Chromosome Location
Chr. 16: 56643687 - 56644941 on Build GRCh38
UniGene ID
Hs.679600
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF348999.2 - 3 166

View Full Product Details