Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001099666.1 | NP_001093136.1 | 2, 3 | 285 | |
XM_005251976.4 | XP_005252033.1 | 2, 3 | 375 | ||
XM_005251977.4 | XP_005252034.1 | 2, 3 | 349 | ||
XM_005251980.4 | XP_005252037.1 | 2, 3 | 335 | ||
XM_005251981.4 | XP_005252038.1 | 2, 3 | 335 | ||
XM_011518640.1 | XP_011516942.1 | 2, 3 | 369 | ||
GenBank mRNA | BC053622.1 | - | 2, 3 | 267 |