Gene Transcripts

Gene Symbol
MCC
Entrez Gene ID
4163
Gene Name
mutated in colorectal cancers
Gene Aliases
MCC1
Chromosome Location
Chr. 5: 113022099 - 113488830 on Build GRCh38
UniGene ID
Hs.593171
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085377.1 NP_001078846.1 1 390
GenBank mRNA AK128596.1 - 1 390
CF887436.1 - 1 384

View Full Product Details