Gene Transcripts

Gene Symbol
REP15
Entrez Gene ID
387849
Gene Name
RAB15 effector protein
Gene Aliases
RAB15EP
Chromosome Location
Chr. 12: 27696495 - 27697633 on Build GRCh38
UniGene ID
Hs.269836
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029874.1 NP_001025045.1 1 186
GenBank mRNA AY662682.1 - 1 121
BC140921.1 - 1 162 9056733

View Full Product Details