Gene Transcripts
- Gene Symbol
- TPTEP1
- Entrez Gene ID
- 387590
- Gene Name
- transmembrane phosphatase with tensin homology pseudogene 1
- Gene Aliases
- psiTPTE22
- Chromosome Location
- Chr. 22: 16601911 - 16648830 on Build GRCh38
- UniGene ID
- Hs.474116
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK095032.1 |
- |
1 |
428 |
|
AK226145.1 |
- |
5 |
1151 |
|
View Full Product Details