Gene Transcripts
- Gene Symbol
- PARP3
- Entrez Gene ID
- 10039
- Gene Name
- poly(ADP-ribose) polymerase family member 3
- Gene Aliases
- ADPRT3, ADPRTL2, ADPRTL3, ARTD3, IRT1, PADPRT-3
- Chromosome Location
- Chr. 3: 51942282 - 51948867 on Build GRCh38
- UniGene ID
- Hs.271742
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001003931.3 |
NP_001003931.3
|
2, 3 |
597 |
|
NM_005485.5 |
NP_005476.4
|
2, 3 |
602 |
|
XM_005264779.4 |
XP_005264836.2
|
3, 4 |
742 |
|
XM_017005490.1 |
XP_016860979.1
|
3, 4 |
729 |
|
GenBank mRNA |
AB208803.1 |
- |
2, 3 |
482 |
|
AF083068.1 |
- |
2, 3 |
321 |
|
AY126341.1 |
- |
2, 3 |
432 |
|
BC014260.2 |
- |
1, 2 |
203 |
4763951 |
BG818516.1 |
- |
3, 4 |
681 |
|
BI554046.1 |
- |
2, 3 |
536 |
|
Y16836.1 |
- |
2, 3 |
556 |
|
View Full Product Details