Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005856.2 | NP_005847.1 | 3 | 360 | |
XM_006715631.3 | XP_006715694.1 | 5 | 683 | ||
XM_017011666.1 | XP_016867155.1 | 3 | 724 | ||
GenBank mRNA | AJ001016.1 | - | 3 | 328 | |
AK128765.1 | - | 1 | 1690 | ||
AK311871.1 | - | 3 | 299 | ||
AY265459.1 | - | 3 | 299 | ||
BC022304.1 | - | 3 | 327 | 4717934 | |
BC053852.1 | - | 3 | 326 | 6197363 | |
BP347072.1 | - | 3 | 360 |