Gene Transcripts

Gene Symbol
PRR22
Entrez Gene ID
163154
Gene Name
proline rich 22
Gene Aliases
Chromosome Location
Chr. 19: 5782960 - 5784765 on Build GRCh38
UniGene ID
Hs.631838
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134316.1 NP_001127788.1 3 1125
GenBank mRNA BC008985.1 - 1 1373
BC023278.1 - 1 1373 4213314

View Full Product Details