Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000398.6 | NP_000389.1 | 7 | 669 | |
NM_001129819.2 | NP_001123291.1 | 7 | 913 | ||
NM_001171660.1 | NP_001165131.1 | 7 | 809 | ||
NM_001171661.1 | NP_001165132.1 | 8 | 729 | ||
NM_007326.4 | NP_015565.1 | 7 | 668 | ||
GenBank mRNA | AF361370.1 | - | 7 | 604 | |
AJ010116.1 | - | 7 | 582 | ||
AJ010117.1 | - | 7 | 582 | ||
AJ010118.1 | - | 7 | 582 | ||
AJ310899.1 | - | 4 | 286 | ||
AJ310900.1 | - | 4 | 286 | ||
AK124528.1 | - | 4 | 2719 | ||
AK302204.1 | - | 7 | 808 | ||
AY421733.1 | - | 2 | 132 | ||
BC004821.1 | - | 7 | 613 | 3450773 | |
BM564146.1 | - | 8 | 719 | ||
BT009821.1 | - | 7 | 585 | ||
BX334220.2 | - | 7 | 670 | ||
BX366240.2 | - | 7 | 669 | ||
CR456435.1 | - | 7 | 591 | ||
M16461.1 | - | 6 | 566 | ||
M16462.1 | - | 6 | 504 | ||
Y09501.1 | - | 7 | 604 |