Gene Transcripts
- Gene Symbol
- PLD3
- Entrez Gene ID
- 23646
- Gene Name
- phospholipase D family member 3
- Gene Aliases
- AD19, HU-K4, HUK4
- Chromosome Location
- Chr. 19: 40348395 - 40378490 on Build GRCh38
- UniGene ID
- Hs.257008
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001031696.3 |
NP_001026866.1
|
5, 6 |
651 |
|
NM_001291311.1 |
NP_001278240.1
|
5, 6 |
585 |
|
NM_012268.3 |
NP_036400.2
|
5, 6 |
875 |
|
XM_005258704.1 |
XP_005258761.1
|
5, 6 |
796 |
|
XM_005258707.4 |
XP_005258764.1
|
5, 6 |
578 |
|
XM_005258708.3 |
XP_005258765.1
|
4, 5 |
471 |
|
XM_005258709.4 |
XP_005258766.1
|
4, 5 |
459 |
|
XM_005258710.4 |
XP_005258767.1
|
4, 5 |
500 |
|
XM_006723122.1 |
XP_006723185.1
|
5, 6 |
963 |
|
XM_011526692.1 |
XP_011524994.1
|
5, 6 |
958 |
|
XM_011526693.1 |
XP_011524995.1
|
5, 6 |
678 |
|
XM_017026546.1 |
XP_016882035.1
|
6, 7 |
894 |
|
XM_017026547.1 |
XP_016882036.1
|
5, 6 |
462 |
|
XM_017026548.1 |
XP_016882037.1
|
5, 6 |
613 |
|
XM_017026549.1 |
XP_016882038.1
|
5, 6 |
716 |
|
GenBank mRNA |
AK293690.1 |
- |
4, 5 |
376 |
|
BC000553.1 |
- |
1, 2 |
121 |
|
BC036327.1 |
- |
5, 6 |
376 |
5189261 |
BC096820.1 |
- |
5, 6 |
574 |
30558682 |
BP304863.1 |
- |
4, 5 |
391 |
|
U60644.1 |
- |
5, 6 |
581 |
|
View Full Product Details