Gene Transcripts

Gene Symbol
TIPARP
Entrez Gene ID
25976
Gene Name
TCDD inducible poly(ADP-ribose) polymerase
Gene Aliases
ARTD14, PARP7, pART14
Chromosome Location
Chr. 3: 156674416 - 156706770 on Build GRCh38
UniGene ID
Hs.744050
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184717.1 NP_001171646.1 3 1465
NM_001184718.1 NP_001171647.1 3 1030
NM_015508.4 NP_056323.2 3 1380
GenBank mRNA AK097785.1 - 3 1209
AK291261.1 - 3 1215
AL080156.1 - 2 66
BC034397.1 - 3 1030
BC050350.1 - 3 1187 5264807
BX537965.1 - 3 1207
CR749647.1 - 3 1466

View Full Product Details