Gene Transcripts

Gene Symbol
SIRPB2
Entrez Gene ID
284759
Gene Name
signal regulatory protein beta 2
Gene Aliases
PTPN1L, PTPNS1L3, dJ776F14.2
Chromosome Location
Chr. 20: 1474591 - 1491762 on Build GRCh38
UniGene ID
Hs.721685
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001122962.1 NP_001116434.1 3 883
NM_001134836.1 NP_001128308.1 3 589
XM_005260708.4 XP_005260765.1 3 692
XM_005260709.3 XP_005260766.1 3 555
XM_011529221.2 XP_011527523.1 3 1479
XM_011529223.2 XP_011527525.1 4 927
XM_011529224.2 XP_011527526.1 3 371
XM_011529225.2 XP_011527527.1 2 292
GenBank mRNA AK095499.1 - 3 692
AK294440.1 - 4 578
AK297072.1 - 3 483
AY424277.1 - 3 361

View Full Product Details