Gene Transcripts

Gene Symbol
MANSC1
Entrez Gene ID
54682
Gene Name
MANSC domain containing 1
Gene Aliases
9130403P13Rik, LOH12CR3
Chromosome Location
Chr. 12: 12329263 - 12350541 on Build GRCh38
UniGene ID
Hs.591145
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018050.2 NP_060520.2 4 1236
XM_017019531.1 XP_016875020.1 5 1440
GenBank mRNA AK001160.1 - 5 1141
AK023622.1 - 4 1236
AK298681.1 - 2 871
BC032998.2 - 4 1253 4824116

View Full Product Details