Gene Transcripts

Gene Symbol
OTUD4
Entrez Gene ID
54726
Gene Name
OTU deubiquitinase 4
Gene Aliases
DUBA6, HIN1, HSHIN1
Chromosome Location
Chr. 4: 145133650 - 145180019 on Build GRCh38
UniGene ID
Hs.270851
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001102653.1 NP_001096123.1 3, 4 245
NM_017493.6 NP_059963.1 2, 3 143
XM_005263079.4 XP_005263136.1 3, 4 348
XM_011532040.2 XP_011530342.2 3, 4 349
XM_011532041.2 XP_011530343.2 3, 4 349
XM_011532042.2 XP_011530344.1 3, 4 270
GenBank mRNA AK302581.1 - 3, 4 207
BC118572.1 - 2, 3 139 40036249
BC118653.1 - 2, 3 139 40036248
DB446926.2 - 3, 4 247
X68242.1 - 3, 4 234

View Full Product Details