Gene Transcripts
- Gene Symbol
- PRODH
- Entrez Gene ID
- 5625
- Gene Name
- proline dehydrogenase 1
- Gene Aliases
- HSPOX2, PIG6, POX, PRODH1, PRODH2, TP53I6
- Chromosome Location
- Chr. 22: 18912774 - 18936553 on Build GRCh38
- UniGene ID
- Hs.517352
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001195226.1 |
NP_001182155.1
|
3, 4 |
301 |
|
NM_016335.4 |
NP_057419.4
|
4, 5 |
716 |
|
GenBank mRNA |
AB209472.1 |
- |
3, 4 |
367 |
|
BC036534.1 |
- |
3, 4 |
525 |
|
BC068260.1 |
- |
3, 4 |
517 |
|
BC094736.1 |
- |
3, 4 |
346 |
|
BC118597.1 |
- |
3, 4 |
301 |
40108132 |
BC121809.1 |
- |
3, 4 |
301 |
40108133 |
KJ534920.1 |
- |
3, 4 |
512 |
|
U79754.1 |
- |
4, 5 |
716 |
|
- Gene Symbol
- LOC102724788
- Entrez Gene ID
- 102724788
- Gene Name
- proline dehydrogenase 1, mitochondrial
- Gene Aliases
- Chromosome Location
- Chr. 22|NT_187389: 138086 - 161619 on Build GRCh38
- Species
- Homo sapiens
View Full Product Details