Gene Transcripts

Gene Symbol
KLHL14
Entrez Gene ID
57565
Gene Name
kelch like family member 14
Gene Aliases
Chromosome Location
Chr. 18: 32672673 - 32773011 on Build GRCh38
UniGene ID
Hs.446164
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020805.2 NP_065856.1 7 1946
GenBank mRNA AB037805.1 - 7 1946
AK294996.1 - 6 1184

View Full Product Details