Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001033561.1 | NP_001028733.1 | 4 | 1101 | |
NM_001290131.1 | NP_001277060.1 | 4 | 1101 | ||
NM_020889.2 | NP_065940.1 | 4 | 1101 | ||
XM_005258015.3 | XP_005258072.1 | 3 | 390 | ||
XM_005258016.3 | XP_005258073.1 | 3 | 410 | ||
XM_011525078.2 | XP_011523380.1 | 3 | 702 | ||
XM_011525079.2 | XP_011523381.1 | 4 | 1084 | ||
XM_011525082.2 | XP_011523384.1 | 4 | 1084 | ||
XM_011525083.2 | XP_011523385.1 | 4 | 1084 | ||
XM_017024903.1 | XP_016880392.1 | 3 | 1149 | ||
XM_017024904.1 | XP_016880393.1 | 3 | 669 | ||
GenBank mRNA | AK160370.1 | - | 4 | 3128 | |
AK291420.1 | - | 4 | 573 | ||
AK294056.1 | - | 3 | 530 | ||
AY030283.1 | - | 4 | 1103 | ||
BC001657.1 | - | 2 | 318 | ||
BC110882.1 | - | 4 | 918 | 6157238 | |
BC121043.1 | - | 4 | 603 | 40117907 | |
BC121044.1 | - | 4 | 603 | 40117911 |