Gene Transcripts
- Gene Symbol
- RTN1
- Entrez Gene ID
- 6252
- Gene Name
- reticulon 1
- Gene Aliases
- NSP
- Chromosome Location
- Chr. 14: 59595976 - 59871288 on Build GRCh38
- UniGene ID
- Hs.368626
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021136.2 |
NP_066959.1
|
8, 9 |
2487 |
|
NM_206852.2 |
NP_996734.1
|
6, 7 |
882 |
|
XM_017021578.1 |
XP_016877067.1
|
6, 7 |
771 |
|
GenBank mRNA |
AK128113.1 |
- |
3, 4 |
2347 |
|
AK290534.1 |
- |
6, 7 |
882 |
|
AK297492.1 |
- |
7, 8 |
789 |
|
BC000314.2 |
- |
6, 7 |
667 |
|
BC003003.1 |
- |
6, 7 |
667 |
|
BC090862.1 |
- |
8, 9 |
2402 |
6179803 |
BC111694.1 |
- |
9, 10 |
2177 |
40035553 |
BX449967.2 |
- |
6, 7 |
870 |
|
CR457030.1 |
- |
6, 7 |
574 |
|
L10333.1 |
- |
8, 9 |
2400 |
|
L10334.1 |
- |
6, 7 |
1308 |
|
L10335.1 |
- |
6, 7 |
602 |
|
View Full Product Details