Gene Transcripts
- Gene Symbol
- SIAH1
- Entrez Gene ID
- 6477
- Gene Name
- siah E3 ubiquitin protein ligase 1
- Gene Aliases
- SIAH1A
- Chromosome Location
- Chr. 16: 48360531 - 48448440 on Build GRCh38
- UniGene ID
- Hs.706828
- Species
- Homo sapiens
- Gene Symbol
- LOC100507577
- Entrez Gene ID
- 100507577
- Gene Name
- uncharacterized LOC100507577
- Gene Aliases
- Chromosome Location
- Chr. 16: 48355685 - 48362999 on Build GRCh38
- UniGene ID
- Hs.744137
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040677.1 |
|
2 |
1710 |
|
GenBank mRNA |
AK056051.1 |
- |
2 |
1709 |
|
View Full Product Details