Gene Transcripts
- Gene Symbol
- SPTBN2
- Entrez Gene ID
- 6712
- Gene Name
- spectrin beta, non-erythrocytic 2
- Gene Aliases
- GTRAP41, SCA5, SCAR14
- Chromosome Location
- Chr. 11: 66682496 - 66729361 on Build GRCh38
- UniGene ID
- Hs.26915
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006946.2 |
NP_008877.1
|
5, 6 |
654 |
|
XM_005274192.4 |
XP_005274249.1
|
6, 7 |
1316 |
|
XM_005274193.3 |
XP_005274250.1
|
5, 6 |
1090 |
|
XM_006718669.3 |
XP_006718732.1
|
4, 5 |
3003 |
|
XM_006718671.3 |
XP_006718734.1
|
6, 7 |
716 |
|
XM_011545216.2 |
XP_011543518.1
|
4, 5 |
3013 |
|
XM_011545217.2 |
XP_011543519.1
|
4, 5 |
3013 |
|
XM_017018174.1 |
XP_016873663.1
|
6, 7 |
1408 |
|
XM_017018175.1 |
XP_016873664.1
|
6, 7 |
1037 |
|
XM_017018176.1 |
XP_016873665.1
|
6, 7 |
757 |
|
XM_017018177.1 |
XP_016873666.1
|
5, 6 |
946 |
|
XM_017018178.1 |
XP_016873667.1
|
5, 6 |
629 |
|
GenBank mRNA |
AB008567.1 |
- |
5, 6 |
654 |
|
BC139788.1 |
- |
5, 6 |
625 |
|
View Full Product Details