Gene Transcripts
- Gene Symbol
- KAZALD1
- Entrez Gene ID
- 81621
- Gene Name
- Kazal type serine peptidase inhibitor domain 1
- Gene Aliases
- BONO1, FKSG28, FKSG40, IGFBP-rP10
- Chromosome Location
- Chr. 10: 101061141 - 101068141 on Build GRCh38
- UniGene ID
- Hs.534859
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001319303.1 |
NP_001306232.1
|
4, 5 |
1803 |
|
NM_030929.4 |
NP_112191.2
|
4, 5 |
1748 |
|
NR_135067.1 |
|
3, 4 |
1186 |
|
XM_017016714.1 |
XP_016872203.1
|
3, 4 |
827 |
|
XM_017016715.1 |
XP_016872204.1
|
3, 4 |
827 |
|
GenBank mRNA |
AK172862.1 |
- |
4, 5 |
999 |
|
AK172864.1 |
- |
4, 5 |
1802 |
|
AK225949.1 |
- |
4, 5 |
1000 |
|
AY359087.1 |
- |
4, 5 |
1004 |
|
BC007758.2 |
- |
4, 5 |
1149 |
4122366 |
BG034107.1 |
- |
|
12 |
|
BI766345.1 |
- |
3, 4 |
459 |
|
CX871488.1 |
- |
3, 4 |
477 |
|
View Full Product Details