Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001146160.1 | NP_001139632.1 | 3 | 149 | |
NM_001317889.1 | NP_001304818.1 | 5 | 265 | ||
NM_001317890.1 | NP_001304819.1 | 3 | 248 | ||
NM_032026.3 | NP_114415.1 | 5 | 265 | ||
NR_027427.1 | 5 | 265 | |||
XM_006716666.3 | XP_006716729.1 | 5 | 285 | ||
XM_006716667.3 | XP_006716730.1 | 5 | 330 | ||
XM_006716668.3 | XP_006716731.1 | 5 | 343 | ||
XM_006716669.3 | XP_006716732.1 | 3 | 227 | ||
XM_006716670.3 | XP_006716733.1 | 4 | 186 | ||
XM_006716671.3 | XP_006716734.1 | 4 | 264 | ||
XM_011517331.2 | XP_011515633.1 | 4 | 181 | ||
XM_017013895.1 | XP_016869384.1 | 5 | 275 | ||
XM_017013896.1 | XP_016869385.1 | 5 | 330 | ||
XM_017013897.1 | XP_016869386.1 | 4 | 251 | ||
XM_017013898.1 | XP_016869387.1 | 4 | 188 | ||
XM_017013899.1 | XP_016869388.1 | 5 | 261 | ||
GenBank mRNA | AF212250.1 | - | 5 | 248 | |
AK312204.1 | - | 5 | 261 | ||
AY071865.1 | - | 3 | 138 | ||
BC064964.1 | - | 5 | 249 | 6141834 | |
BG532054.1 | - | 3 | 238 | ||
BX332564.2 | - | 5 | 247 |