Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033329.2 | NP_201586.1 | 1 | 212 | |
NM_053003.3 | NP_443729.1 | 2 | 602 | ||
XM_011527441.2 | XP_011525743.1 | 2 | 299 | ||
XM_011527442.2 | XP_011525744.1 | 2 | 602 | ||
GenBank mRNA | AF282256.1 | - | 2 | 582 | |
AK095919.1 | - | 2 | 602 | ||
AY358140.1 | - | 2 | 560 | ||
BC035809.1 | - | 2 | 599 | 5755526 |