Gene Transcripts
- Gene Symbol
- BTAF1
- Entrez Gene ID
- 9044
- Gene Name
- B-TFIID TATA-box binding protein associated factor 1
- Gene Aliases
- MOT1, TAF(II)170, TAF172, TAFII170
- Chromosome Location
- Chr. 10: 91923769 - 92030998 on Build GRCh38
- UniGene ID
- Hs.500526
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003972.2 |
NP_003963.1
|
35, 36 |
5181 |
|
XM_011540326.1 |
XP_011538628.1
|
35, 36 |
5280 |
|
XM_011540327.2 |
XP_011538629.1
|
32, 33 |
4946 |
|
XM_011540328.2 |
XP_011538630.1
|
32, 33 |
4774 |
|
XM_017016877.1 |
XP_016872366.1
|
34, 35 |
5243 |
|
XM_017016878.1 |
XP_016872367.1
|
34, 35 |
5073 |
|
XM_017016879.1 |
XP_016872368.1
|
34, 35 |
5197 |
|
XM_017016880.1 |
XP_016872369.1
|
34, 35 |
5201 |
|
XM_017016881.1 |
XP_016872370.1
|
33, 34 |
5050 |
|
GenBank mRNA |
AB208843.1 |
- |
23, 24 |
3575 |
|
AF038362.1 |
- |
35, 36 |
5200 |
|
AJ001017.2 |
- |
35, 36 |
5095 |
|
AK303554.1 |
- |
13, 14 |
1858 |
|
BC029930.1 |
- |
11, 12 |
1576 |
|
BC112201.1 |
- |
35, 36 |
5165 |
8327669 |
View Full Product Details