Gene Transcripts

Gene Symbol
LPAR2
Entrez Gene ID
9170
Gene Name
lysophosphatidic acid receptor 2
Gene Aliases
EDG-4, EDG4, LPA-2, LPA2
Chromosome Location
Chr. 19: 19623655 - 19628930 on Build GRCh38
UniGene ID
Hs.122575
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004720.5 NP_004711.2 3 1101
XM_011528421.2 XP_011526723.1 3 1465
XM_017027468.1 XP_016882957.1 5 1396
XM_017027469.1 XP_016882958.1 4 1199
GenBank mRNA AF011466.1 - 3 1046
AF197929.1 - 3 1080
AF233092.1 - 2 968
AK123043.1 - 6 1865
AK316573.1 - 3 1062
AY322548.1 - 2 962
BC025695.1 - 3 1141 5209267

View Full Product Details