Gene Transcripts
- Gene Symbol
- WTAP
- Entrez Gene ID
- 9589
- Gene Name
- Wilms tumor 1 associated protein
- Gene Aliases
- Mum2
- Chromosome Location
- Chr. 6: 159726680 - 159756319 on Build GRCh38
- UniGene ID
- Hs.446091
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001270531.1 |
NP_001257460.1
|
5, 6 |
477 |
|
NM_001270532.1 |
NP_001257461.1
|
5, 6 |
609 |
|
NM_001270533.1 |
NP_001257462.1
|
5, 6 |
477 |
|
NM_004906.4 |
NP_004897.2
|
5, 6 |
609 |
|
NM_152857.2 |
NP_690596.1
|
5, 6 |
477 |
|
NM_152858.2 |
NP_690597.1
|
5, 6 |
609 |
|
XM_017011514.1 |
XP_016867003.1
|
5, 6 |
447 |
|
XM_017011515.1 |
XP_016867004.1
|
3, 4 |
709 |
|
XM_017011516.1 |
XP_016867005.1
|
5, 6 |
447 |
|
GenBank mRNA |
AF277190.1 |
- |
1, 2 |
49 |
|
AF374416.1 |
- |
5, 6 |
490 |
|
AJ276706.1 |
- |
4, 5 |
257 |
|
AK127822.1 |
- |
5, 6 |
488 |
|
AK290854.1 |
- |
5, 6 |
490 |
|
BC000383.2 |
- |
5, 6 |
468 |
|
BC004432.1 |
- |
5, 6 |
468 |
|
BC069192.1 |
- |
5, 6 |
445 |
4653831 |
BG702882.1 |
- |
5, 6 |
481 |
|
BQ948913.1 |
- |
5, 6 |
415 |
|
BX387163.2 |
- |
5, 6 |
459 |
|
CN284477.1 |
- |
5, 6 |
439 |
|
D14661.1 |
- |
5, 6 |
389 |
|
DA561768.1 |
- |
5, 6 |
483 |
|
View Full Product Details