The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105206.2 | NP_001098676.2 | 3 - 4 | 700 | 87 |
NM_001105207.2 | NP_001098677.2 | 3 - 4 | 717 | 87 | |
NM_002290.4 | NP_002281.3 | 3 - 4 | 700 | 87 | |
XM_005266983.3 | XP_005267040.2 | 3 - 4 | 573 | 87 | |
XM_005266984.3 | XP_005267041.2 | 3 - 4 | 570 | 87 | |
XM_017010854.1 | XP_016866343.1 | 3 - 4 | 570 | 87 | |
GenBank mRNA | AB210027.1 | - | 3 - 4 | 552 | 87 |
AK304401.1 | - | 3 - 4 | 523 | 87 | |
BC026237.1 | - | 3 - 4 | 532 | 87 | |
BC066552.1 | - | 3 - 4 | 713 | 87 | |
S78569.1 | - | 3 - 4 | 492 | 87 | |
X91171.1 | - | 3 - 4 | 585 | 87 |