Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYBPC1
Entrez Gene ID
4604
Gene Name
myosin binding protein C, slow type
Gene Aliases
LCCS4, MYBPCC, MYBPCS
Chromosome Location
Chr.12: 101594849 - 101695841 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001254718.1 NP_001241647.1 9 - 10 901 75
NM_001254719.1 NP_001241648.1 9 - 10 901 75
NM_001254720.1 NP_001241649.1 8 - 9 865 75
NM_001254721.1 NP_001241650.1 8 - 9 844 75
NM_001254722.1 NP_001241651.1 7 - 8 823 75
NM_001254723.1 NP_001241652.1 8 - 9 862 75
NM_002465.3 NP_002456.2 11 - 12 976 75
NM_206819.2 NP_996555.1 11 - 12 976 75
NM_206820.2 NP_996556.1 9 - 10 901 75
NM_206821.2 NP_996557.1 9 - 10 901 75
XM_005268876.4 XP_005268933.1 10 - 11 910 75
XM_006719405.3 XP_006719468.1 11 - 12 837 75
XM_006719406.3 XP_006719469.1 11 - 12 837 75
XM_006719407.3 XP_006719470.1 10 - 11 910 75
XM_006719408.3 XP_006719471.1 11 - 12 837 75
XM_006719409.3 XP_006719472.1 9 - 10 863 75
XM_006719410.3 XP_006719473.1 11 - 12 837 75
XM_006719411.3 XP_006719474.1 9 - 10 863 75
XM_017019315.1 XP_016874804.1 10 - 11 780 75
XM_017019316.1 XP_016874805.1 11 - 12 837 75
XM_017019317.1 XP_016874806.1 9 - 10 863 75
XM_017019318.1 XP_016874807.1 10 - 11 780 75
XM_017019319.1 XP_016874808.1 8 - 9 806 75
XM_017019320.1 XP_016874809.1 7 - 8 905 75
XM_017019321.1 XP_016874810.1 8 - 9 806 75
XM_017019322.1 XP_016874811.1 6 - 7 848 75
GenBank mRNA AK056587.1 - 10 - 11 900 75
AK292976.1 - 11 - 12 936 75
AK295727.1 - 7 - 8 783 75
AK296681.1 - 9 - 10 865 75
AK303401.1 - 8 - 9 822 75
AL831993.2 - 11 - 12 911 75
AL832000.2 - 11 - 12 911 75
AL832332.2 - 11 - 12 911 75
AL832350.2 - 11 - 12 911 75
AL832354.2 - 11 - 12 913 75
AL832456.2 - 11 - 12 911 75
AL834249.2 - 11 - 12 911 75
BC015187.1 - 11 - 12 936 75
BC065720.1 - 7 - 8 786 75
BC092418.1 - 9 - 10 848 75
BC117217.1 - 7 - 8 758 75
BC143495.1 - 8 - 9 779 75
BC143502.1 - 7 - 8 758 75
BC143503.1 - 9 - 10 836 75
BC143504.1 - 8 - 9 800 75
BX649073.2 - 12 - 13 1007 75
CD614901.1 - 6 - 7 601 75
X73114.1 - 9 - 10 842 75

View Full Product Details