Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RGS10
Entrez Gene ID
6001
Gene Name
regulator of G-protein signaling 10
Gene Aliases
-
Chromosome Location
Chr.10: 119499813 - 119542752 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002925.3 NP_002916.1 1 - 2 68 82
XM_017016492.1 XP_016871981.1 1 - 2 116 82
GenBank mRNA AF045229.2 - 1 - 2 48 82
AL547312.3 - 1 - 2 68 82
AV707839.1 - 1 - 2 68 82

View Full Product Details