Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- RAD51D
- Entrez Gene ID
-
5892
- Gene Name
- RAD51 paralog D
- Gene Aliases
-
BROVCA4, R51H3, RAD51L3, TRAD
- Chromosome Location
-
Chr.17: 35099792 - 35119869 on Build GRCh38
- Gene Symbol
- RAD51L3-RFFL
- Entrez Gene ID
-
100529207
- Gene Name
- RAD51L3-RFFL readthrough
- Gene Aliases
-
-
- Chromosome Location
-
Chr.17: 35009112 - 35121522 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_037714.1 |
|
2 - 3 |
332 |
69 |
GenBank mRNA |
AK097811.1 |
- |
2 - 3 |
303 |
69 |
View Full Product Details