Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NR2C1
Entrez Gene ID
7181
Gene Name
nuclear receptor subfamily 2 group C member 1
Gene Aliases
TR2
Chromosome Location
Chr.12: 95020229 - 95073628 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032287.2 NP_001027458.1 3 - 4 545 63
NM_001127362.1 NP_001120834.1 3 - 4 545 63
NM_003297.3 NP_003288.2 3 - 4 545 63
XM_005269133.2 XP_005269190.1 3 - 4 532 63
XM_006719584.2 XP_006719647.1 3 - 4 532 63
XM_006719585.2 XP_006719648.1 4 - 5 570 63
XM_011538716.2 XP_011537018.1 3 - 4 532 63
XM_011538717.2 XP_011537019.1 3 - 4 532 63
GenBank mRNA AB307707.1 - 2 - 3 289 63
AK291319.1 - 3 - 4 509 63
BC026074.1 - 3 - 4 491 63
BC040141.1 - 3 - 4 497 63
DB266298.1 - 3 - 4 545 63
JQ308177.1 - 2 - 3 289 63
JQ308178.1 - 2 - 3 289 63
M21985.1 - 3 - 4 415 63
M29959.1 - 3 - 4 415 63
M29960.1 - 3 - 4 345 63

View Full Product Details