Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRKCE
Entrez Gene ID
5581
Gene Name
protein kinase C epsilon
Gene Aliases
PKCE, nPKC-epsilon
Chromosome Location
Chr.2: 45651315 - 46187990 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005400.2 NP_005391.1 5 - 6 895 91
XM_005264428.1 XP_005264485.1 6 - 7 924 91
XM_005264431.3 XP_005264488.1 5 - 6 1484 91
XM_011532971.2 XP_011531273.1 7 - 8 1548 91
XM_011532975.2 XP_011531277.1 9 - 10 1036 91
XM_011532978.1 XP_011531280.1 6 - 7 528 91
XM_011532980.2 XP_011531282.1 6 - 7 565 91
XM_011532981.2 XP_011531283.1 4 - 5 1688 91
XM_011532982.1 XP_011531284.1 3 - 4 700 91
XM_017004486.1 XP_016859975.1 7 - 8 1590 91
XM_017004487.1 XP_016859976.1 7 - 8 637 91
XM_017004488.1 XP_016859977.1 6 - 7 1372 91
XM_017004489.1 XP_016859978.1 10 - 11 977 91
XM_017004490.1 XP_016859979.1 6 - 7 1477 91
XM_017004491.1 XP_016859980.1 5 - 6 528 91
XM_017004492.1 XP_016859981.1 5 - 6 1484 91
GenBank mRNA AK313842.1 - 5 - 6 794 91
BC109033.2 - 5 - 6 833 91
BC109034.2 - 5 - 6 833 91
X65293.1 - 5 - 6 713 91

View Full Product Details