Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAPRE2
Entrez Gene ID
10982
Gene Name
microtubule associated protein RP/EB family member 2
Gene Aliases
CSCSC2, EB1, EB2, RP1
Chromosome Location
Chr.18: 34976928 - 35143470 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143826.2 NP_001137298.1 5 - 6 700 86
NM_001143827.2 NP_001137299.1 5 - 6 846 86
NM_001256420.1 NP_001243349.1 3 - 4 660 86
NM_014268.3 NP_055083.1 4 - 5 788 86
NR_046177.1 4 - 5 603 86
GenBank mRNA AB016823.1 - 4 - 5 723 86
AK294833.1 - 5 - 6 790 86
AK296251.1 - 4 - 5 603 86
AK315766.1 - 4 - 5 741 86
BC007318.2 - 4 - 5 731 86
BT020086.1 - 4 - 5 608 86
CR536545.1 - 4 - 5 608 86
X94232.1 - 4 - 5 748 86

View Full Product Details