The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004272.4 | NP_004263.1 | 6 - 7 | 1747 | 75 |
GenBank mRNA | AF093262.1 | - | 5 - 6 | 685 | 75 |
AK223471.1 | - | 6 - 7 | 1009 | 75 | |
AK312481.1 | - | 6 - 7 | 1012 | 75 | |
AY189940.1 | - | 2 - 3 | 183 | 75 | |
BC015502.1 | - | 6 - 7 | 962 | 75 | |
BC047438.1 | - | 6 - 7 | 1747 | 75 | |
BT009846.1 | - | 5 - 6 | 685 | 75 | |
Y17829.1 | - | 6 - 7 | 760 | 75 |