The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004771.3 | NP_004762.2 | 1 - 2 | 137 | 144 |
GenBank mRNA | Y12779.1 | - | 1 - 2 | 137 | 144 |