The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143970.1 | NP_001137442.1 | 5 - 6 | 1524 | 70 |
NM_001143971.1 | NP_001137443.1 | 5 - 6 | 1448 | 70 | |
NM_004824.3 | NP_004815.3 | 5 - 6 | 1675 | 70 | |
NR_026590.1 | 7 - 8 | 1630 | 70 | ||
XM_011514997.2 | XP_011513299.1 | 5 - 6 | 1604 | 70 | |
GenBank mRNA | AF081258.1 | - | 7 - 8 | 1630 | 70 |
AF081259.1 | - | 7 - 8 | 1630 | 70 | |
AK291601.1 | - | 6 - 7 | 1438 | 70 | |
AK296985.1 | - | 5 - 6 | 1524 | 70 | |
AL050164.1 | - | 1 - 2 | 136 | 70 | |
BC061516.1 | - | 4 - 5 | 745 | 70 | |
BC108725.1 | - | 5 - 6 | 1686 | 70 | |
BC119682.2 | - | 4 - 5 | 671 | 70 |