Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM2
Entrez Gene ID
23670
Gene Name
transmembrane protein 2
Gene Aliases
-
Chromosome Location
Chr.9: 71683366 - 71769532 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135820.1 NP_001129292.1 9 - 10 2401 93
NM_013390.2 NP_037522.1 10 - 11 2590 93
XM_005251869.4 XP_005251926.1 10 - 11 2223 93
GenBank mRNA AB037833.2 - 10 - 11 2183 93
AF137030.1 - 10 - 11 2198 93
AK075370.1 - 3 - 4 318 93
BC140773.1 - 9 - 10 1903 93
BC144510.1 - 10 - 11 2058 93
BC146780.1 - 10 - 11 2183 93
BC171782.1 - 9 - 10 1904 93

View Full Product Details