Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLITRK3
Entrez Gene ID
22865
Gene Name
SLIT and NTRK like family member 3
Gene Aliases
-
Chromosome Location
Chr.3: 165186720 - 165197109 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014926.3 NP_055741.2 1 - 2 259 92
GenBank mRNA AB020655.1 - 1 - 2 88 92
BC114621.1 - 1 - 2 73 92
DA123399.1 - 1 - 2 261 92

View Full Product Details