The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001139488.1 | NP_001132960.1 | 10 - 11 | 1798 | 90 |
NM_015376.2 | NP_056191.1 | 9 - 10 | 1356 | 90 | |
NM_170672.2 | NP_733772.1 | 11 - 12 | 1711 | 90 | |
XM_005264246.3 | XP_005264303.1 | 10 - 11 | 2138 | 90 | |
XM_011532746.2 | XP_011531048.1 | 11 - 12 | 1585 | 90 | |
XM_011532747.2 | XP_011531049.1 | 10 - 11 | 1600 | 90 | |
XM_011532748.2 | XP_011531050.2 | 10 - 11 | 1446 | 90 | |
XM_017003759.1 | XP_016859248.1 | 12 - 13 | 3061 | 90 | |
XM_017003760.1 | XP_016859249.1 | 11 - 12 | 3533 | 90 | |
XM_017003761.1 | XP_016859250.1 | 10 - 11 | 1463 | 90 | |
XM_017003762.1 | XP_016859251.1 | 11 - 12 | 1712 | 90 | |
XM_017003763.1 | XP_016859252.1 | 10 - 11 | 1442 | 90 | |
GenBank mRNA | AB020653.1 | - | 9 - 10 | 1356 | 90 |
AK292983.1 | - | 11 - 12 | 1622 | 90 | |
BC027849.1 | - | 11 - 12 | 1711 | 90 |