The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016184.3 | NP_057268.1 | 2 - 3 | 440 | 114 |
XM_017019382.1 | XP_016874871.1 | 3 - 4 | 361 | 114 | |
GenBank mRNA | AF067800.1 | - | 2 - 3 | 461 | 114 |
AF109146.1 | - | 2 - 3 | 287 | 114 | |
AF328684.1 | - | 2 - 3 | 432 | 114 | |
AJ133532.1 | - | 2 - 3 | 434 | 114 | |
BC074822.2 | - | 2 - 3 | 215 | 114 | |
BC074874.2 | - | 2 - 3 | 215 | 114 | |
BC117439.1 | - | 2 - 3 | 335 | 114 | |
BC117441.1 | - | 2 - 3 | 335 | 114 |