Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIRT6
Entrez Gene ID
51548
Gene Name
sirtuin 6
Gene Aliases
SIR2L6
Chromosome Location
Chr.19: 4174109 - 4182604 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193285.2 NP_001180214.1 1 - 2 136 79
NM_001321059.1 NP_001307988.1 1 - 2 136 79
NM_001321060.1 NP_001307989.1 1 - 2 136 79
NM_001321063.1 NP_001307992.1 1 - 2 136 79
NM_016539.3 NP_057623.2 1 - 2 136 79
GenBank mRNA AF233396.1 - 1 - 2 131 79
AK074810.1 - 1 - 2 99 79
AK225567.1 - 1 - 2 104 79
AK301247.1 - 1 - 2 99 79
AK315048.1 - 1 - 2 100 79
BC005026.1 - 1 - 2 74 79
BC028220.1 - 1 - 2 82 79
CN364175.1 - 1 - 2 79 79
CN364177.1 - 1 - 2 138 80
CR457200.1 - 1 - 2 71 79

View Full Product Details